Stampa…
Wall bracket for eyewash bottles
Fornitore: ORKLA CARE
7202
CEDE7202EA 0 EUR
CEDE7202
Wall bracket for eyewash bottles
Ganci da parete
Two holders in green hard plastics intended for storage of two Cederroth eyewash bottles (500 ml), making it easy to find in an emergency situation.
Holders are fastened on a metal plate, which can be mounted on any vertical surface example on a wall.
Also serves as an opening device for the eyewash bottles: Bottle is automatically opened when removed from the holder.
Afterglowing sign with instructions for use is included.