Stampa…
Catalase Assay Kit
Fornitore: Cayman Chemical
707002-480
CAYM707002-480EA 1230 EUR
CAYM707002-480
Catalase Assay Kit
Saggi Saggi enzimatici
For the measurement of catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, and cell lysates.
  • Assay 40 samples in duplicate
  • Measure catalase activity down to 2 U/ml
  • Plate-based colorimetric measurement (540 nm)
Catalase is an ubiquitous antioxidant enzyme that is present in most aerobic cells. It is involved in the detoxification of H2O2, a ROS that is a toxic product of both normal aerobic metabolism and pathogenic ROS production. Cayman’s Catalase Assay utilises the peroxidatic function of catalase for determination of enzyme activity.