Stampa…

α-Idrossi-o-tolil β-D-glucopiranoside

Fornitore: Cayman Chemical

Synonyms: 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, D(-)-Salicin, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol, D(−)-Salicin

17357-10 17357-100 17357-25 17357-50
CAYM17357-10EA 124 EUR
CAYM17357-10 CAYM17357-100 CAYM17357-25 CAYM17357-50
α-Idrossi-o-tolil β-D-glucopiranoside
α-Idrossi-o-tolil β-D-glucopiranoside

Salicin is an alcoholic β-glucoside derived from willow bark that produces anti-inflammatory (including analgesic and antipyretic) effects very similar to that of aspirin. It acts as a non-selective COX inhibitor, exhibiting IC50 values >100 µM for COX-1 and COX-2.

Formula: C₁₃H₁₈O₇
MDL Number: MFCD00006590
CAS Number: 138-52-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR