Stampa…

Anti-GPR43 Rabbit Polyclonal Antibody

Fornitore: Cayman Chemical
15727-1
CAYM15727-1EA 682 EUR
CAYM15727-1
Anti-GPR43 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: GPR43
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR