Stampa…

Anti-Grx1 Goat Polyclonal Antibody (Biotin)

Fornitore: Cayman Chemical
11546-1
CAYM11546-1EA 661 EUR
CAYM11546-1
Anti-Grx1 Goat Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: Grx1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: E. Coli
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR