Stampa…

3-(4,5-Dimetiltiazol-2-il)-2,5-difeniltetrazolio bromuro (MTT)

Fornitore: Cayman Chemical
Attenzione

Synonyms: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide

10009591-1 10009591-5
CAYM10009591-1EA 93 EUR
CAYM10009591-1 CAYM10009591-5
3-(4,5-Dimetiltiazol-2-il)-2,5-difeniltetrazolio bromuro (MTT)
3-(4,5-Dimetiltiazol-2-il)-2,5-difeniltetrazolio bromuro (MTT)
Formula: C₁₈H₁₆N₅BrS
Peso molecolare: 414.33 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00011964
CAS Number: 298-93-1
EINECS: 206-069-5

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR