Stampa…
Cooler, Powerbox® Plus
Fornitore: CAMPING GAZ
2000024955
CAMP2000024955EA 104 EUR
CAMP2000024955
Cooler, Powerbox® Plus
Scatole Sistema di raffreddamento da banco
The Powerbox® Plus electric cooler is packed with 30% more premium insulation than its predecessor and keep content 20 °C below the ambient temperature and the fan emits a maximum of 39 dB.
  • Thicker and improved PU insulation
  • Firmly locking lid
  • Noise reduced fan
Cooler Powerbox® Plus has antimicrobial lining that comes from a chemical additive injected into the plastic within the liner.