Stampa…
CAMAG® TLC Visualizer 3 Documentation System
Fornitore: CAMAG
022.9850
CAMA022.9850EA 0 EUR
CAMA022.9850
CAMAG® TLC Visualizer 3 Documentation System
Sistemi di documentazione fotografica
CAMAG® TLC Visualizer 3 documentation system with 12 mm lens, without visionCATS software. Suited for object formats up to about 21×28 cm (20×20 cm TLC plates).
  • Maximum illumination homogeneity in white light, UV 366 and UV 254 nm
  • High-performance industrial camera with CMOS image sensor
  • Powered by visionCATS HPTLC analysis software
  • Various image enhancement tools for the detection of even the faintest zones
  • Side by side display of tracks from different plates
  • Image-based evaluation
  • Detector: Industrial camera with CMOS image sensor
  • Connections: USB 3.0