Stampa…

o-Cresolphthalein, Sigma-Aldrich®

Fornitore: SIGMA ALDRICH MICROSCOPY

Synonyms: 3,3-Bis(4-hydroxy-3-methylphenyl)isobenzofuran-1(3H)-one

MQ200

C85778-10G
C85778-10GEA 42.7 EUR
C85778-10G
o-Cresolphthalein, Sigma-Aldrich®
o-Cresolphthalein
Formula: C₂₂H₁₈O₄
Peso molecolare: 346.39 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005912
CAS Number: 596-27-0
EINECS: 209-881-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=654afb7b32a4c01d8bae3e3622e1393974f93945&vid=4e2ad92a16fd4eab054ce7445a0f34369f040bbe&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR