Stampa…

Guanidinio cloruro solution_SC 6 M, GenAR® Biotechnology, Macron Fine Chemicals™

Fornitore: Avantor Performance Materials
Attenzione

Synonyms: Guanidina cloridrato

C0563-10EA 951.28 EUR
C0563-10 0563-20 MACR0563-26
Guanidinio cloruro solution_SC 6 M, GenAR® Biotechnology, Macron Fine Chemicals™
Guanidinio cloruro
Formula: NH₂C(=NH)NH₂·HCl
Peso molecolare: 95.53 g/mol
Punto di fusione: 185
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (clear, colorless liquid) Passes Test
Cyanide (CN) Passes Test
Heavy Metals (as Pb) ≤10 ppm
Identification (by FTIR) Passes Test
ACS - Iron (Fe) ≤3 ppm
Molarity (by titrimetry) 5.8 - 6.2
pH (1:10 dilution) 4.5 - 6.5
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR