Stampa…

Anti-CK17 Rabbit Polyclonal Antibody

Fornitore: Bioworld Technology
BS6649
BWRLBS6649EA 341 EUR
BWRLBS6649
Anti-CK17 Rabbit Polyclonal Antibody
Anticorpi
Recombinant full length Human Cytokeratin 17.

Cytokeratin 17 pAb detects endogenous levels of Cytokeratin 17 protein.

Type: Primary
Antigen: CK17
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR