Stampa…

Anti-p-EGFR Rabbit Polyclonal Antibody [clone: Y845 ]

Fornitore: Bioworld Technology
BS6349
BWRLBS6349EA 379 EUR
BWRLBS6349
Anti-p-EGFR Rabbit Polyclonal Antibody [clone: Y845 ]
Anticorpi
Synthetic phosphopeptide derived from human EGFR around the phosphorylation site of Tyrosine 845.

p-EGFR (Y845) pAb detects endogenous levels of EGFR protein only when phosphorylated at Tyr845 (removal of the signal peptide).

Type: Primary
Antigen: p-EGFR
Clonality: Polyclonal
Clone: Y845
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR