Stampa…

Anti-SNX1 Rabbit Polyclonal Antibody

Fornitore: Bioworld Technology
BS5940
BWRLBS5940EA 262 EUR
BWRLBS5940
Anti-SNX1 Rabbit Polyclonal Antibody
Anticorpi
Synthetic peptide, corresponding to amino acids 458-503 of Human SNX1.

SNX1 pAb detects endogenous levels of SNX1 protein.

Type: Primary
Antigen: SNX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR