Stampa…

Anti-NCAM-L1 Rabbit Polyclonal Antibody [clone: R1177]

Fornitore: Bioworld Technology
BS3723
BWRLBS3723EA 262 EUR
BWRLBS3723
Anti-NCAM-L1 Rabbit Polyclonal Antibody [clone: R1177]
Anticorpi
Synthetic peptide, corresponding to amino acids 1150-1200 of Human NCAM-L1.

NCAM-L1 (R1177) pAb detects endogenous levels of NCAM-L1 protein.

Type: Primary
Antigen: NCAM-L1
Clonality: Polyclonal
Clone: R1177
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR