Stampa…

Anti-ND1 Rabbit Polyclonal Antibody [clone: V208]

Fornitore: Bioworld Technology
BS3338
BWRLBS3338EA 262 EUR
BWRLBS3338
Anti-ND1 Rabbit Polyclonal Antibody [clone: V208]
Anticorpi
Synthetic peptide, corresponding to amino acids 171-230 of Human ND1.

ND1 (V208) pAb detects endogenous levels of ND1 protein.

Type: Primary
Antigen: ND1
Clonality: Polyclonal
Clone: V208
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Specifiche

Simbolo dell'antigene ND1
Environmentally Preferable
Tipo anticorpo Primary
Clonalità Policlonale
Clone V208
Reattività Human
Ospite Rabbit
Isotipo
Western blot Yes
ImmunoChimica Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR