Stampa…

Anti-c-Fms Rabbit Polyclonal Antibody [clone: Q718]

Fornitore: Bioworld Technology
BS3245
BWRLBS3245EA 262 EUR
BWRLBS3245
Anti-c-Fms Rabbit Polyclonal Antibody [clone: Q718]
Anticorpi
Synthetic peptide, corresponding to amino acids 691-740 of Human c-Fms.

c-Fms (Q718) pAb detects endogenous levels of c-Fms protein.

Type: Primary
Antigen: c-Fms
Clonality: Polyclonal
Clone: Q718
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR