Stampa…

Anti-TUSC3 Rabbit Polyclonal Antibody [clone: H153]

Fornitore: Bioworld Technology
BS2048
BWRLBS2048EA 262 EUR
BWRLBS2048
Anti-TUSC3 Rabbit Polyclonal Antibody [clone: H153]
Anticorpi
Synthetic peptide, corresponding to amino acids 122-171 of Human TUSC3.

TUSC3 (H153) pAb detects endogenous levels of TUSC3 protein.

Type: Primary
Antigen: TUSC3
Clonality: Polyclonal
Clone: H153
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR