Stampa…

Anti-Ephrin-A1 Rabbit Polyclonal Antibody [clone: K98]

Fornitore: Bioworld Technology
BS1950
BWRLBS1950EA 262 EUR
BWRLBS1950
Anti-Ephrin-A1 Rabbit Polyclonal Antibody [clone: K98]
Anticorpi
Synthetic peptide, corresponding to amino acids 70-120 of Human Ephrin-A1.

Ephrin-A1 (K98) pAb detects endogenous levels of Ephrin-A1 protein.

Type: Primary
Antigen: Ephrin-A1
Clonality: Polyclonal
Clone: K98
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR