Stampa…
Drum pump for mineral oils
Fornitore: BURKLE
5763-0100
BURK5763-0100EA 3280 EUR
BURK5763-0100
Drum pump for mineral oils
Pompe Pompe a tamburo
Barrel pump is used for light viscous mineral oil products such as diesel, fuel oil, hydraulic fluid, machinery, motor oil and many more.
  • Pump tube made of aluminium
  • Delivery pump height 19 mWS
Informazioni per l'ordine: Supplied with 230 V motor, hose connection Alu (1"), (2 m) PVC fabric hose (1"), hose clips (1"), Alu pump nozzle, PP barrel screw joint, medium-contacting parts made of aluminium and AISI 316 Ti.