Stampa…
Scoops
Fornitore: BURKLE
5370-1000 5370-2000 5370-0120 5370-0450 5370-1500
BURK5370-1000EA 56.4 EUR
BURK5370-1000 BURK5370-2000 BURK5370-0120 BURK5370-0450 BURK5370-1500
Scoops
Palette
Sterilisable scoops made of stainless steel AISI 304 (1.4301).
  • Robust
  • Autoclavable up to 121 °C for max. 20 min.
  • Suitable for food contact