Stampa…
Instrument dishes
Fornitore: BURKLE
4230-2070 4230-2071 4230-2180 4230-2230 4230-2100 4230-2101 4230-2231 4230-2550 4230-2050
BURK4230-2070EA 16.2 EUR
BURK4230-2070 BURK4230-2071 BURK4230-2180 BURK4230-2230 BURK4230-2100 BURK4230-2101 BURK4230-2231 BURK4230-2550 BURK4230-2050
Instrument dishes
Vassoi/Piatti/Contenitori Vassoi per strumenti
Trays for various uses. In laboratories, doctors’ surgeries, hospitals, pharmacies, the foodstuffs industry - wherever there is a need for clean storage of instruments, equipment, goods and precision parts.
  • Melamine, white
  • Tray without lid
Lid available as accessory.