Stampa…
Diaphragm vacuum pumps, V-300
Fornitore: BUCHI
11V300000 11V300010 11V300020 11V300100 11V300110 11V300120 11V300200 11V300210 11V300220 11V300300 11V300310 11V300320
BUCH11V300000EA 0 EUR
BUCH11V300000 BUCH11V300010 BUCH11V300020 BUCH11V300100 BUCH11V300110 BUCH11V300120 BUCH11V300200 BUCH11V300210 BUCH11V300220 BUCH11V300300 BUCH11V300310 BUCH11V300320
Diaphragm vacuum pumps, V-300
Pompe Pompe da vuoto a membrana
These V-300 PTFE diaphragm vacuum pumps impresses with its very silent and economical operation. These pumps have been operated at a flow rate of 1,8 m³/h and an ultimate vacuum of 5 mbar.
  • High chemical resistant due to the use of high quality materials
  • Very low noise emission
  • Easy visual detection of condensate and particles due to transparent front panel
The speed-controlled operation in conjunction with the optional Interface I-300 / I-300 Pro increases the lifetime of the pumps. In combination with the Interface I-300 / I-300 Pro an optionalMore Product Information