Stampa…
Melting point apparatus, M-565
Fornitore: BUCHI
051998 11058004
621-0044EA 0 EUR
621-0044 BUCH11058004
Melting point apparatus, M-565
Apparecchi per punto di fusione
M-565 is an advanced, automated instrument with video capability for professional quality control in a regulated environment. Offers all the features above but with the automated determination of melting and boiling points. The product is available as a 'Professional set' with or without the M-569 sample loader - please see below for details.
  • Real time video displayed in 6× magnification
  • Video replay function with variable speed
  • Exclusive optional accessories including MeltingPoint Monitor software and IQ/OQ documentation
  • Reproducible results with M-569
  • M-5