Stampa…
Anti-IgA Goat Polyclonal Antibody (CF™ 594)
Fornitore: Biotium
20429-50UL 20429-500UL
BTIU20429-50ULEA 121 EUR
BTIU20429-50UL BTIU20429-500UL
Anti-IgA Goat Polyclonal Antibody (CF™ 594)
Anticorpi
Goat anti-human IgA reacts specifically with IgA heavy chains (α chains) and not with immunoglobulin light chains. To minimize crossreactivity, it has been adsorbed human IgG prior to conjugation. This conjugate is labeled with CF™594 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Deep-red fluorescent CF™594 (Ex/Em 593/614 nm) is spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugate, and has excellent photostability.
Alexa Fluor® and Texas Red® are registered trademarks of ThermoMore Product Information