Stampa…
Pasteur pipettes, disposable
Fornitore: Brand
747750 747755 747760 747765 747770 747775
BRND747750EA 60.8 EUR
BRND747750 612-1714 BRND747760 BRND747765 612-1715 BRND747775
Pasteur pipettes, disposable
Pipette Pipette Pasteur/di trasferimento
LDPE.
  • Excellent at reproducing number of drops per mililitre
  • Ideal for aliquots
  • Pasteur pipettes can be filled and deep frozen, or changed into a closed vessel by heat-sealing the tip
  • The integrated pipetting bulb depresses easily, minimising fatigue from frequent pipetting
  • Resistant to gas or gamma radiation sterilisation