Stampa…
Matracci da filtrazione con connettore
Fornitore: Brand
73714 73719 73724 73729
BRND73714EA 54 EUR
BRND73714 BRND73719 BRND73724 BRND73729
Matracci da filtrazione con connettore
Fiasche Matracci filtranti
Borosilicate glass 3.3.
  • Erlenmeyer shape
  • Ground socket to accommodate vacuum tubing of 15 to 18 mm external diameter (e.g. 6×5 or 8×5 mm)
  • Fulfil the regulations of the 'Equipment and product safety regulations'
For vacuum use. Filtration flasks with side arm socket have not only made work in preparation and analytical laboratories easier and simpler, but have also reduced accident risk.
Certificazioni: DIN 12476 and ISO 6556