Stampa…
Clips for conical standard ground joints
Fornitore: Brand
55646
BRND55646EA 8.9 EUR
BRND55646
Clips for conical standard ground joints
Morsetti Morsetti per giunti NS
POM. Will not scratch glass surfaces.
  • Excellent resilience and stability
  • Outstanding chemical resistance
  • Operating temperature up to 150 °C