Stampa…
Pipette tray for drawer
Fornitore: Brand
28305
BRND28305EA 54.7 EUR
BRND28305
Pipette tray for drawer
Vassoi/Piatti/Contenitori Vassoi per pipette
Pipette tray made of PVC.
  • Four compartments
  • For approx. 30 graduated pipettes (1 to 20 ml)