Stampa…
Self-laminating labels, MiniMark
Fornitore: Brady
710113
BRDY710113EA 438.89 EUR
BRDY710113
Self-laminating labels, MiniMark
Etichette
The MiniMark self-laminating wire and cable markers are designed to adhere to highly curved surfaces, wires and cables. They feature a white printable area with clear tail that wraps around marker. The clear tail protects the text, providing superior resistance to abrasion, dirt, oils, solvents, water and excessive handling.