Stampa…
Polyester Autoclave and Cryogenic Labels for M710
Fornitore: Brady
311984 311981 311990 311991 311982 311978
BRDY311984EA 52.61 EUR
BRDY311984 BRDY311981 BRDY311990 BRDY311991 BRDY311982 BRDY311978
Polyester Autoclave and Cryogenic Labels for M710
Etichette
FreezerBondz™ material is a low-profile label used on frozen surfaces or on room-temperature surfaces that will eventually be stored in Liquid Nitrogen (-196°C).
  • Can be applied to frozen surfaces, including glass and polypropylene stored in liquid nitrogen
  • Good print smudge resistance and solvent resistance
  • High/low-temperature performance range of −196 to +100 °C
  • Matte finish
Identify frosted, frozen, or room temperature vials (with overlap). Opaque enough to cover legally protected patient information for compliance with HIPAA requirements. IdealMore Product Information