Stampa…
ISO safety signs - Fire alarm flashing light
Fornitore: Brady
311199 311200 311201 311202 311203 311204 311214 311215 311216 311217 311218 311219 311205 311206 311207 311208 311209 311210 311211 311212 311213 311220 311221 311222 311223 311224 311225 311226 311227 311228 311229
BRDY311199EA 3.37 EUR
BRDY311199 BRDY311200 BRDY311201 BRDY311202 BRDY311203 BRDY311204 BRDY311214 BRDY311215 BRDY311216 BRDY311217 BRDY311218 BRDY311219 BRDY311205 BRDY311206 BRDY311207 BRDY311208 BRDY311209 BRDY311210 BRDY311211 BRDY311212 BRDY311213 BRDY311220 BRDY311221 BRDY311222 BRDY311223 BRDY311224 BRDY311225 BRDY311226 BRDY311227 BRDY311228 BRDY311229
ISO safety signs - Fire alarm flashing light
Segnali
When working in a loud production environment, ear protection is necessary to reduce noise exposure. But can employees still hear the fire alarm?
  • Increase safety using internationally recognised signs
  • Compliant with ISO 7010/ISO 3864
  • Designs available in multiple top selling materials
To solve this issue, fire alarm flashing lights have been developed. ISO recognised the need to clearly indicate the location of fire alarm flashing lights. Production areas usually have many flashing lights which could also warn about the active status of a machine, orMore Product Information