Stampa…
BradyJet J4000-EU-BWSPWID Label Printer, Colour, with Product and Wire ID Software
Fornitore: Brady
310389
BRDY310389EA 3491.76 EUR
BRDY310389
BradyJet J4000-EU-BWSPWID Label Printer, Colour, with Product and Wire ID Software
Stampanti Label Printers
This industry inkjet workhorse creates up to 5000 impactful full-colour labels per day up to 203,20 mm wide on continuous or die-cut material.
  • Full colour
  • Long-lasting
  • 5000 labels per day
Ideal for a wide variety of applications in safety, including lean/5S, lockout tagout, instructional or safety chemical labelling for handling, storage or transport, as well as lab settings. Contains the colour options you need to create ANSI, GHS, ISO, BS5609 and arc flash compliant labels. Streamline storage by only needing to stock white label material and printMore Product Information
Visualizza i codici
Specifiche
Environmentally Preferable
Requisiti informatici PC
Interfacce USB; ethernet
Materiali supportati Continuous and die-cut
Modello J4000-EU-BWSPWID
Alimentazione AC adaptor
Stampante Full Colour
Metodo di stampa Inkjet
Risoluzione di stampa 4800 dpi
Velocità di stampa 5000 max. labels/day
Larghezza di stampa 209,55 mm (max.)