Stampa…
Self-Laminating Vinyl Wrap Around Labels for M710 and BMP71
Fornitore: Brady
174596 173353 174651 173344 174572 173399 174533 173337 174593 173411 174598 173362 174640 174603 173332 173316 173378 173335 174604 173429 174597 174534 173403 173334 173372 174599 174531 173345 174605 174535 174574 174532 174660
BRDY174596EA 59.75 EUR
BRDY174596 BRDY173353 BRDY174651 BRDY173344 BRDY174572 BRDY173399 BRDY174533 BRDY173337 BRDY174593 BRDY173411 BRDY174598 BRDY173362 BRDY174640 BRDY174603 BRDY173332 BRDY173316 BRDY173378 BRDY173335 BRDY174604 BRDY173429 BRDY174597 BRDY174534 BRDY173403 BRDY173334 BRDY173372 BRDY174599 BRDY174531 BRDY173345 BRDY174605 BRDY174535 BRDY174574 BRDY174532 BRDY174660
Self-Laminating Vinyl Wrap Around Labels for M710 and BMP71
Etichette
Self-laminating vinyl (B-427) is an excellent material for wire and cable identification. This material has good clarity and conformability, and is self-extinguishing.
  • Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • Service temperature of -40 to 158 °F (-40 to 70 °C)
  • Matte finish
Ideal for data and telecommunications labelling, electrical labelling, laboratory labelling, and wire and cable labellingMore Product Information