Stampa…
Labels and tape for BBP®35 and BBP®37 thermal transfer benchtop printers
Fornitore: Brady
139950 145298 145300 145292 145293 143579 143580 145295 145294 143584 143581 145296 139949 139948 139947 139946 139945 139944 139943 139942 139941 139940 139939 139938 139937 139936 139935 139934 139932 139929 139928 143578 145297 143582
BRDY139950EA 324.66 EUR
BRDY139950 BRDY145298 BRDY145300 BRDY145292 BRDY145293 BRDY143579 BRDY143580 BRDY145295 BRDY145294 BRDY143584 BRDY143581 BRDY145296 BRDY139949 BRDY139948 BRDY139947 BRDY139946 BRDY139945 BRDY139944 BRDY139943 BRDY139942 BRDY139941 BRDY139940 BRDY139939 BRDY139938 BRDY139937 BRDY139936 BRDY139935 BRDY139934 BRDY139932 BRDY139929 BRDY139928 BRDY143578 BRDY145297 BRDY143582
Labels and tape for BBP®35 and BBP®37 thermal transfer benchtop printers
Etichette
A variety of polyester and specialty materials for thermal transfer printing in wide format rolls designed for shape cutting multiple labels out of a single continuous roll.
B-423
This material is ideal for smooth-surface indoor labeling such as datacom equipment, patch panels, face plates, data ports and general industrial identification.
B-424
White matte paper labels with permanent rubber based adhesive for general purpose labelling.
B-428
Metallised polyester tape is ideal for indoor product labeling, asset tracking and general ID on smoother surfaces with a higher temperature limit.
B-432