Stampa…
Ribbons, multicolour, 'colour and cut' for GlobalMark® label markers
Fornitore: Brady
076739 076741 076751 076771 076743 076745 076747 076762 076763 076790 076791 076775
116-0417EA 309.17 EUR
116-0417 BRDY076741 BRDY076751 BRDY076771 BRDY076743 BRDY076745 BRDY076747 BRDY076762 BRDY076763 BRDY076790 BRDY076791 BRDY076775
Ribbons, multicolour, 'colour and cut' for GlobalMark® label markers
Nastri per stampanti
Super-tough smear proof GlobalMark® multicolour and colour & cut ribbons assure the printed text lasts for years outdoors and in an industrial environment.
  • Assorted contains red, blue, green, cyan, yellow, magenta, black colour ribbons
2 and 4-colour 'panelled' ribbons are used to print labels and signs containing more than 1 print colour.
Width: 105 mm
Informazioni per l'ordine: Ribbons are supplied self-contained in cartridges that drop straight in to the Globalmark with a 'click'.