Stampa…
Labels for LaserTab® laser printer, type B-747
Fornitore: Brady
029718 029837 029727 029696 029738
BRDY029718EA 654.35 EUR
BRDY029718 BRDY029837 BRDY029727 BRDY029696 BRDY029738
Labels for LaserTab® laser printer, type B-747
Etichette
Blank, printable polyester labels for Laser Printers.
  • Designed to be printed with a Laser or InkJet printer
  • Available blank or pre-printed with a template or header
  • Supplied on 203,2×254 mm or A4 209,8×297,2 mm sheets
This material is a UL recognised component when printed with designated laser printer toners. See UL file MH17154 for specific details.