Stampa…
Thermal transfer printer labels, type B-490, FreezerBondz
Fornitore: Brady
011264 011265 011263 011266 011267 114203 114202 114204 114200 114201
BRDY011264EA 224.14 EUR
BRDY011264 BRDY011265 BRDY011263 BRDY011266 BRDY011267 BRDY114203 BRDY114202 BRDY114204 BRDY114200 BRDY114201
Thermal transfer printer labels, type B-490, FreezerBondz
Etichette
FreezerBondz thermal transfer printers labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen. These labels are on a 1" core, a 'smaller core' than the majority of labels. Recommended printer for these is the BBP11-34L printer.
  • Good print smudge resistance
  • Solvent resistance
  • Good high and low temperature performance
  • Widely used in liquid nitrogen and autoclave applications
Freezerbondz™ printable labels for thermal transfer printers. Freezerbondz™ labels can be applied to frozen surfaces including glassMore Product Information