Stampa…
Sharps disposal container, Medibox®
Fornitore: B. Braun
9193627 9193618 9193424 9193639
BRAU9193627EA 332 EUR
BRAU9193627 BRAU9193618 BRAU9193424 BRAU9193639
Sharps disposal container, Medibox®
Bidoni Contenitori per lame
Medibox® containers are suitable for easy and safe disposal of all kinds of medical sharps. Consist of diverse ports for disposal without contact. Comply with international standards.
  • Impact and puncture resistant
  • Freestanding, ergonomic design for easy handling
  • Handling instructions by self-explaining pictograms
  • Large opening to ease the insertion of various medical sharps
  • Overfill warning by maximum fill line and visible content level
  • Irreversible final lock mechanism
Certificazioni: EN ISO 23907