Stampa…
b.safe Screw Caps
Fornitore: Bohlender
M575-01
BOHLM575-01EA 8 EUR
BOHLM575-01
b.safe Screw Caps
Ghiere Tappi per bottiglie
Screw cap made of PP with handy knurl and internal PTFE/silicone gasket. After assembly, the product is only exposed to PTFE.
  • Made of PP
  • Very good chemical resistance
  • Equipped with PTFE/silicone seal
Screw caps are designed for closing unused connections on a b.safe Distributor for flushing bottles, or also to close eluent bottles during the flushing process.