Stampa…

Screw caps with aperture

Fornitore: Bohlender
H995-45 H983-01 H984-02 H983-02 H983-03 H995-18 H983-04 H984-05 H984-04 H984-03 H983-05 H995-32 H984-01 H995-14
BOHLH995-45EA 19.5 EUR
BOHLH995-45 BOHLH983-01 BOHLH984-02 BOHLH983-02 BOHLH983-03 BOHLH995-18 BOHLH983-04 BOHLH984-05 BOHLH984-04 BOHLH984-03 BOHLH983-05 BOHLH995-32 BOHLH984-01 BOHLH995-14
Screw caps with aperture
Ghiere Tappi per bottiglie
PTFE/Glass fibre, PBTP or PPS.

  • Suitable gasket rings and septums are available separately

Screw cap with handy knurl, suitable for GL threads, with aperture. Joining piece between tubes with flange and tubes with GL thread.

PTFE/Glass fibre
Temperature resistance: −200 to +250 °C
Universal chemical resistance

PBPT
Temperature resistance: from −45 to +140 °C
Good chemical resistance

PPS
Temperature resistance: from −20 to +250 °C
Universal chemical resistance
Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR