Stampa…
Seals for threaded connection caps, double sided, BOLA
Fornitore: Bohlender
H977-08 H977-16 H977-18 H977-36 H977-32 H977-28 H977-20
BOHLH977-08EA 5.9 EUR
BOHLH977-08 BOHLH977-16 BOHLH977-18 BOHLH977-36 BOHLH977-32 BOHLH977-28 BOHLH977-20
Seals for threaded connection caps, double sided, BOLA
Ghiere Tappi per bottiglie
Silicone ring with double-sided PTFE washer. After assembly, the product is only exposed to PTFE.
  • Temperature resistance: from –60 to +230 °C
  • Universal chemical resistance