Stampa…
Magnetic stirring bar retrievers, PTFE, BOLA
Fornitore: Bohlender
C372-12 C372-06 C372-04 C372-02 C372-18 C372-08 C372-10 C371-16
442-9839EA 17 EUR
442-9839 BOHLC372-06 BOHLC372-04 BOHLC372-02 BOHLC372-18 BOHLC372-08 BOHLC372-10 BOHLC371-16
Magnetic stirring bar retrievers, PTFE, BOLA
Dispositivi agitatori Accessori di agitazione
PTFE-encapsulated stirring bar retrievers with extra strong permanent magnet (Neodym). For the removal of stirring bars from aggressive liquids. Especially useful for big, heavy stirring bars up to 400 g.
  • Very good chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +250 °C)