Stampa…
Retort clamp, MAXI
Fornitore: BOCHEM
5567
BOCH5567EA 444 EUR
BOCH5567
Retort clamp, MAXI
Morsetti Clamp per uso generico
Retort clamp made of chromium plated steel.
  • Fibreglass covered
  • 400 mm total length