Stampa…
Double bossheads
Fornitore: BOCHEM
5350 5352 5356 5357 5353 5354
BOCH5350EA 22.4 EUR
BOCH5350 BOCH5352 BOCH5356 BOCH5357 BOCH5353 BOCH5354
Double bossheads
Morsetti Morsetti
Double bossheads, either made of stainless steel or aluminium.
  • Angle: 180 + 90°
  • Different screw types available