Stampa…
Anti-KIT Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KIT/982]
Fornitore: Biotium
BNUM0982-50
BNUM0982-50EA 417 EUR
BNUM0982-50
Anti-KIT Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KIT/982]
Anticorpi
This MAb recognizes a protein of 145 kDa, identified as CD117/p145kit. It is found on a wide variety of tumor cells including follicular and papillary carcinoma of thyroid, adenocarcinomas from endometrium, lung, ovary, pancreas, and breast as well as malignant melanoma, endodermal sinus tumor, and small cell carcinoma. However, anti-CD117 has been particularly useful in differentiating gastrointestinal stromal tumors from Kaposi's sarcoma, tumors of smooth muscle origin, fibromatosis, and neural tumors of the GI tract. Anti-CD117 is also useful in recognizing myeloblasts in bone marrowMore Product Information