Stampa…
Anti-BCL6 Mouse Monoclonal Antibody (CF594) [clone: BCL6/850]
Fornitore: Biotium
BNC940850-500 BNC940850-100
BNC940850-500EA 560 EUR
BNC940850-500 BNC940850-100
Anti-BCL6 Mouse Monoclonal Antibody (CF594) [clone: BCL6/850]
Anticorpi
Recognizes a protein of 95 kDa, which is identified as Bcl-6. Antibody to bcl-6 is helpful in a number of diagnostic settings: (1) In the differential diagnosis of small B-cell lymphoma. Follicular lymphoma will show bcl-6 (and CD10) positivity whereas other small B-cell lymphomas are usually negative. (2) Bcl-6 is an important prognostic marker in diffuse large B-cell lymphomas (DLBCL), where CD10, bcl-6 and MUM1/IRF4 are used to identify germinal center and activated B-cell phenotypes. (3) Bcl-6 can be valuable in distinguishing classical Hodgkin lymphoma from nodular lymphocyteMore Product Information