Stampa…
Anti-KRT18 Mouse Monoclonal Antibody (CF594) [clone: KRT18/835]
Fornitore: Biotium
BNC940835-500 BNC940835-100
BNC940835-500EA 560 EUR
BNC940835-500 BNC940835-100
Anti-KRT18 Mouse Monoclonal Antibody (CF594) [clone: KRT18/835]
Anticorpi
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It is reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine andMore Product Information