Stampa…
Anti-KRT17 Mouse Monoclonal Antibody (CF594) [clone: E3]
Fornitore: Biotium
BNC940158-500 BNC940158-100
BNC940158-500EA 560 EUR
BNC940158-500 BNC940158-100
Anti-KRT17 Mouse Monoclonal Antibody (CF594) [clone: E3]
Anticorpi
Cytokeratin 17 (CK17) is normally expressed in the basal cells of complex epithelia but not in stratified or simple epithelia. Antibody to CK17 is an excellent tool to distinguish myoepithelial cells from luminal epithelium of various glands such as mammary, sweat and salivary. CK17 is expressed in epithelial cells of various origins, such as bronchial epithelial cells and skin appendages. It may be considered as anepithelial stem cellmarker because CK17 Ab marks basal cell differentiation. CK17 is expressed in SCLC much higher than in LADC. Eighty-five percent of the triple negative breastMore Product Information