Stampa…
Anti-CNN1 Mouse Monoclonal Antibody (CF488A) [clone: CNN1/832]
Fornitore: Biotium
BNC880832-500 BNC880832-100
BNC880832-500EA 560 EUR
BNC880832-500 BNC880832-100
Anti-CNN1 Mouse Monoclonal Antibody (CF488A) [clone: CNN1/832]
Anticorpi
Multiple isoelectric variants of calponin have been identified, however only two molecular weight isoforms exist; a 34 kDa form and a 29 kDa form. Expression of the 29 kDa form, I-calponin, is primarily restricted to muscle of the urogenital tract, whereas the higher molecular weight variant has been demonstrated in vascular and visceral smooth muscle. In Western blotting, this MAb reacts with only the 34 kDa form of calponin in extracts of human aortic medial smooth muscle and is unreactive with fibroblast extracts of cultivated human foreskin. Calponin is a calmodulin, F-actin andMore Product Information