Stampa…
Anti-ERBB2 Mouse Monoclonal Antibody (CF488A) [clone: HRB2/718]
Fornitore: Biotium
BNC880718-500 BNC880718-100
BNC880718-500EA 560 EUR
BNC880718-500 BNC880718-100
Anti-ERBB2 Mouse Monoclonal Antibody (CF488A) [clone: HRB2/718]
Anticorpi
Recognizes a protein of 185 kDa, which is identified as c-erbB-2/HER-2/neu. Its epitope is localized in the extracellular domain. C-erbB-2/HER-2 is a member of the EGFR family. This MAb is specific and shows minimal cross-reaction with other members of the EGFR-family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. c-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary.
Type: Primary